3-Methyl-4-nitroanisole 97%

Synonyms: 1-Methoxy-3-methyl-4-nitrobenzene 5-Methoxy-2-nitrotoluene Methyl 4-nitro-m-tolyl ether

B22348-22 B22348-14 B22348-14 B22348-06 B22348-22 B22348-06
AAB22348-06EA 32.84 USD
CAAAB22348-06 AAB22348-06 AAB22348-22 AAB22348-14 CAAAB22348-22 CAAAB22348-14
3-Methyl-4-nitroanisole 97%
3-Methyl-4-nitroanisole
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 48…50 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007167
CAS Number: 5367-32-8
EINECS: 226-356-9
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR