4-Chloro-m-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Chloro-3-methylaniline
B22234-06
B22234-14
B22234-06
B22234-14
AAB22234-06EA
60.09
USD
AAB22234-06
AAB22234-14
CAAAB22234-06
CAAAB22234-14
4-Chloro-m-toluidine 98%
4-Chloro-m-toluidine
Formula:
C₇H₈ClN MW: 141.6 g/mol Melting Pt: 82…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066332 CAS Number: 7149-75-9 UN: 2239 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...