6-Methoxyquinaldine 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 6-Methoxy-2-methylquinoline

B22199-03 B22199-06 B22199-03 B22199-06
AAB22199-03EA 60.99 USD
AAB22199-03 CAAAB22199-06 CAAAB22199-03 AAB22199-06
6-Methoxyquinaldine 97%
6-Methoxyquinaldine
Formula: C₁₁H₁₁NO
MW: 173.21 g/mol
Melting Pt: 62…66 °C
Density: 0.973
Flash Pt: 47 °C(117 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006761
CAS Number: 1078-28-0
EINECS: 214-080-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR