3-Isobutylpyridine 99%

Supplier: Thermo Scientific Chemicals

B22080-09 B22080-88 B22080-88 B22080-09
AAB22080-09EA 168.99 USD
CAAAB22080-88 AAB22080-09 AAB22080-88 CAAAB22080-09
3-Isobutylpyridine 99%
3-Isobutylpyridine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Density: 0.909
Storage Temperature: Ambient
MDL Number: MFCD02683082
CAS Number: 14159-61-6
EINECS: 238-003-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR