1-Phenylnaphthalene 97%

Supplier: Thermo Scientific Chemicals

B21953-06 B21953-14 B21953-22 B21953-06 B21953-22 B21953-14
AAB21953-06EA 73.89 USD
AAB21953-06 AAB21953-14 CAAAB21953-14 AAB21953-22 CAAAB21953-06 CAAAB21953-22
1-Phenylnaphthalene 97%
1-Phenylnaphthalene
Formula: C₁₆H₁₂
MW: 204.27 g/mol
Boiling Pt: 320 °C
Melting Pt: 45 °C
Density: 1.088
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003983
CAS Number: 605-02-7
EINECS: 210-081-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR