2-Bromo-5-fluorocinnamic acid 98%

Warning

B21784-03 B21784-14 B21784-06 B21784-03 B21784-06 B21784-14
AAB21784-14EA 390.72 USD
AAB21784-14 AAB21784-03 CAAAB21784-03 CAAAB21784-14 CAAAB21784-06 AAB21784-06
2-Bromo-5-fluorocinnamic acid 98%
2-Bromo-5-fluorocinnamic acid
Formula: C₉H₆BrFO₂
MW: 245.05 g/mol
Melting Pt: 188…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00142878
CAS Number: 202865-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR