2,3-Difluoroaniline 98%
|
Warning
|
B21665-06
B21665-03
AAB21665-06EA
123.78
USD
AAB21665-06
AAB21665-03
2,3-Difluoroaniline 98%
2,3-Difluoroaniline
Formula:
C₆H₅F₂N MW: 129.11 g/mol Boiling Pt: 68…69 °C/12 mm Density: 1.274 Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010298 CAS Number: 4519-40-8 EINECS: 224-847-2 UN: 2941 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...