L(-)-Xylose 99%
Supplier: Thermo Scientific Chemicals
B21622-06
B21622-22
B21622-22
B21622-14
B21622-06
B21622-14
AAAB21622-22EA
508.99
USD
CAAAAB21622-22
CAAAB21622-14
AAAB21622-22
AAB21622-06
CAAAB21622-06
AAB21622-14
L(-)-Xylose 99%
L(-)-Xylose
Formula:
C₅H₁₀O₅ MW: 150.13 g/mol Boiling Pt: 331 °C (1013 hPa) Melting Pt: 147…151 °C Density: 1.525 Storage Temperature: Ambient |
MDL Number:
MFCD00151096 CAS Number: 609-06-3 EINECS: 210-174-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...