3-Bromo-4-fluorotoluene 99%

Warning

Synonyms: 1-Bromo-2-fluoro-5-methylbenzene

B21523-14 B21523-06 B21523-06 B21523-14
AAB21523-14EA 148.66 USD
CAAAB21523-06 AAB21523-14 CAAAB21523-14 AAB21523-06
3-Bromo-4-fluorotoluene 99%
3-Bromo-4-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 168…170 °C
Density: 1.507
Flash Pt: 73 °C(163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040827
CAS Number: 452-62-0
EINECS: 207-201-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR