Cyclobutylméthanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cyclobutanemethanol
B21480-14
B21480-22
B21480-22
B21480-14
AAB21480-14EA
206.99
USD
CAAAB21480-14
CAAAAB21480-22
AAB21480-14
AAAB21480-22
Cyclobutylméthanol 99%
Cyclobutylméthanol
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 143…144 °C Density: 0.913 Flash Pt: 39 °C(102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001330 CAS Number: 4415-82-1 EINECS: 224-575-4 UN: 1987 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...