2,4,6-Trifluorobenzyl bromide 97%

Danger

Synonyms: α-Bromo-2,4,6-trifluorotoluene

B21453-03 B21453-06 B21453-03 B21453-06
AAB21453-06EA 431.36 USD
AAB21453-06 CAAAB21453-06 AAB21453-03 CAAAB21453-03
2,4,6-Trifluorobenzyl bromide 97%
2,4,6-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061210
CAS Number: 151411-98-2
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR