Phenyl trifluoroacetate 98%

Warning

Synonyms: Trifluoroacetic phenyl ester

B21304-06 B21304-14 B21304-14 B21304-06
AAB21304-14EA 67.79 USD
AAB21304-14 AAB21304-06 CAAAB21304-14 CAAAB21304-06
Phenyl trifluoroacetate 98%
Phenyl trifluoroacetate
Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 146…147 °C
Melting Pt: –8.5 °C
Density: 1.276
Flash Pt: 52 °C(127 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039232
CAS Number: 500-73-2
EINECS: 207-911-4
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR