2-Methyl-3-nitrobenzotrifluoride 97%

Supplier: Thermo Scientific Chemicals
Danger

B21267-06 B21267-03 B21267-14 B21267-03 B21267-14 B21267-06
AAB21267-03EA 32.48 USD
CAAAB21267-06 AAB21267-03 CAAAB21267-14 CAAAB21267-03 AAB21267-14 AAB21267-06
2-Methyl-3-nitrobenzotrifluoride 97%
2-Methyl-3-nitrobenzotrifluoride
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Boiling Pt: 86 °C/10 mm
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042322
CAS Number: 6656-49-1
EINECS: 229-688-2
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR