2,4,5-Trifluorobenzaldehyde 97%

Warning

B20986-03 B20986-03 B20986-06 B20986-06
AAB20986-06EA 132.4 USD
CAAAB20986-03 CAAAB20986-06 AAB20986-06 AAB20986-03
2,4,5-Trifluorobenzaldehyde 97%
2,4,5-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 174 °C
Density: 1.408
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061196
CAS Number: 165047-24-5
EINECS: 000-000-0
UN: 1989
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR