3-Fluoro-4-(trifluoromethyl)benzyl alcohol 97%

Warning

Synonyms: (3-Fluoro-4-(trifluoromethyl)phenyl)methanol

B20976-06 B20976-06 B20976-03 B20976-03
AAB20976-06EA 286.38 USD
CAAAB20976-03 CAAAB20976-06 AAB20976-06 AAB20976-03
3-Fluoro-4-(trifluoromethyl)benzyl alcohol 97%
3-Fluoro-4-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
MW: 194.13 g/mol
Melting Pt: 28…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00236285
CAS Number: 230295-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR