2,5-Difluorobenzamide 98%

Warning

B20971-03 B20971-06 B20971-14 B20971-03 B20971-14 B20971-06
AAAB20971-14EA 367.41 USD
AAAB20971-14 AAB20971-06 CAAAAB20971-14 CAAAB20971-03 CAAAB20971-06 AAB20971-03
2,5-Difluorobenzamide 98%
2,5-Difluorobenzamide
Formula: C₇H₅F₂NO
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00015548
CAS Number: 85118-03-2
EINECS: 285-655-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR