3,4,5-Trifluorobenzaldehyde 98%

Warning

B20943-06 B20943-03 B20943-06 B20943-03
AAB20943-06EA 175.04 USD
AAB20943-06 AAB20943-03 CAAAB20943-03 CAAAB20943-06
3,4,5-Trifluorobenzaldehyde 98%
3,4,5-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 174 °C
Melting Pt: 24…26 °C
Density: 1.42
Flash Pt: 62 °C(144 °F)
Storage Temperature: Ambient
MDL Number: MFCD00083523
CAS Number: 132123-54-7
EINECS: 000-000-0
UN: 3077
ADR: "9",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR