2-(Trifluoromethyl)phenothiazine 98%

Warning

Synonyms: 2-Trifluoromethyl-10H-phenothiazine2-(Trifluoromethyl)-10H-phenothiazine

B20919-06 B20919-14 B20919-06
AAB20919-06EA 35.99 USD
AAB20919-06 CAAAB20919-14 CAAAB20919-06
2-(Trifluoromethyl)phenothiazine 98%
2-(Trifluoromethyl)phenothiazine
Formula: C₁₃H₈F₃NS
MW: 267.27 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00005018
CAS Number: 92-30-8
EINECS: 202-145-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR