1,1,2,2-Tetramethylcyclopropane 99%

Supplier: Thermo Scientific Chemicals
Danger

B20809-14 B20809-06 B20809-06
AAB20809-14EA 415.14 USD
AAB20809-14 CAAAB20809-06 AAB20809-06
1,1,2,2-Tetramethylcyclopropane 99%
1,1,2,2-Tetramethylcyclopropane
Formula: C₇H₁₄
Boiling Pt: 76 °C
Melting Pt: –80.7 °C
Density: 0.72
Storage Temperature: Ambient
MDL Number: MFCD00060790
CAS Number: 4127-47-3
EINECS: 223-937-9
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR