N,N-Diethylacetamide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid diethylamide

B20762-22 B20762-36 B20762-14 B20762-36 B20762-14 B20762-22
AAB20762-36EA 439.99 USD
AAB20762-36 AAB20762-14 AAB20762-22 CAAAB20762-36 CAAAB20762-14 CAAAB20762-22
N,N-Diethylacetamide 99%
N,N-Diethylacetamide
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 182…186 °C
Density: 0.92
Flash Pt: 76 °C(169 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009047
CAS Number: 685-91-6
EINECS: 211-685-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR