2,4-Bis(trifluoromethyl)benzoic acid 98%

Warning

B20731-06 B20731-03
CAAAB20731-06 CAAAB20731-03
2,4-Bis(trifluoromethyl)benzoic acid 98%
2,4-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042496
CAS Number: 32890-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR