2,6-Dichloro-4-(trifluoromethyl)aniline 97%

Warning

Synonyms: 4-Amino-3,5-dichloro-α,α,α-trifluorotoluene

B20728-06 B20728-03 B20728-06 B20728-03
AAB20728-06EA 84.47 USD
AAB20728-06 CAAAB20728-06 AAB20728-03 CAAAB20728-03
2,6-Dichloro-4-(trifluoromethyl)aniline 97%
2,6-Dichloro-4-(trifluoromethyl)aniline
Formula: C₇H₄Cl₂F₃N
MW: 230.02 g/mol
Boiling Pt: 60…62 °C/1 mm
Melting Pt: 34…37 °C
Density: 1.532
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052918
CAS Number: 24279-39-8
EINECS: 416-430-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR