α'-Bromo-α,α,α-trifluoro-o-xylene 97+%

Danger

Synonyms: 2-(Trifluoromethyl)benzyl bromide 2-(Bromomethyl)benzotrifluoride

B20634-14 B20634-06 B20634-06 B20634-14
AAB20634-06EA 85.45 USD
AAB20634-06 AAB20634-14 CAAAB20634-14 CAAAB20634-06
α'-Bromo-α,α,α-trifluoro-o-xylene 97+%
α'-Bromo-α,α,α-trifluoro-o-xylene
Formula: C₈H₆BrF₃
MW: 239.03 g/mol
Boiling Pt: 71…72 °C/7.5 mm
Melting Pt: 27…30 °C
Density: 1.571
Flash Pt: 83 °C(181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013558
CAS Number: 395-44-8
EINECS: 206-901-7
UN: 1760
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR