(2,4,6-Trimethylphenyl)acetonitrile 98%

Warning

Synonyms: 2,4,6-Trimethylbenzyl cyanide Mesitylacetonitrile

B20534-06 B20534-03 B20534-03 B20534-14 B20534-14 B20534-06
AAB20534-03EA 25.8 USD
CAAAB20534-06 AAB20534-03 CAAAB20534-03 AAB20534-14 CAAAB20534-14 AAB20534-06
(2,4,6-Trimethylphenyl)acetonitrile 98%
(2,4,6-Trimethylphenyl)acetonitrile
Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 120 °C/1 mm
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00013815
CAS Number: 34688-71-6
EINECS: 252-151-9
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR