3-Bromo-5-(trifluoromethyl)aniline 98%

Warning

Synonyms: 3-Amino-5-bromo-α,α,α-trifluorotoluene 5-Bromo-α,α,α-trifluoro-m-toluidine

B20437-06 B20437-06 B20437-03 B20437-03
AAB20437-03EA 41.51 USD
CAAAB20437-03 CAAAB20437-06 AAB20437-03 AAB20437-06
3-Bromo-5-(trifluoromethyl)aniline 98%
3-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 221…223 °C
Density: 1.710
Flash Pt: 107 °C(225 °F)
Storage Temperature: Ambient
MDL Number: MFCD00236205
CAS Number: 54962-75-3
EINECS: 259-412-6
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR