3-Fluoro-5-(trifluoromethyl)benzyl alcohol 97%

Warning

B20387-03 B20387-06 B20387-03 B20387-06
AAB20387-06EA 243.09 USD
AAB20387-06 AAB20387-03 CAAAB20387-03 CAAAB20387-06
3-Fluoro-5-(trifluoromethyl)benzyl alcohol 97%
3-Fluoro-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
MW: 194.13 g/mol
Boiling Pt: 80…82 °C/3 mm
Storage Temperature: Ambient
MDL Number: MFCD00061169
CAS Number: 184970-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR