2,6-Difluorocinnamic acid 98%
|
Warning
|
Synonyms:
(E)-3-(2,6-Difluorophenyl)acrylic acid
B20353-03
B20353-06
CAAAB20353-03
CAAAB20353-06
2,6-Difluorocinnamic acid 98%
2,6-Difluorocinnamic acid
Formula:
C₉H₆F₂O₂ MW: 184.14 g/mol Melting Pt: 124…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010319 CAS Number: 102082-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...