2,3-Difluorobenzoyl chloride 97%

Danger

Synonyms: 2,3-Difluorobenzoylchloride

B20333-06 B20333-06
AAAB20333-06EA 84.13 USD
AAAB20333-06 CAAAAB20333-06
2,3-Difluorobenzoyl chloride 97%
2,3-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 189…190 °C
Density: 1.423
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042499
CAS Number: 18355-73-2
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR