2,3,6-Trifluorobenzaldehyde 97%

Warning

B20319-03 B20319-06 B20319-03 B20319-06
AAB20319-03EA 35.36 USD
AAB20319-03 CAAAB20319-06 CAAAB20319-03 AAB20319-06
2,3,6-Trifluorobenzaldehyde 97%
2,3,6-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 173 °C
Density: 1.425
Flash Pt: 81 °C(177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061195
CAS Number: 104451-70-9
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR