3,4-Difluorocinnamic acid 97%

Warning

B20230-06 B20230-03
CAAAB20230-06 CAAAB20230-03
3,4-Difluorocinnamic acid 97%
3,4-Difluorocinnamic acid
Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00010320
CAS Number: 112897-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR