5-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%

Warning

B20184-06 B20184-03 B20184-03 B20184-06
AAB20184-06EA 195.36 USD
CAAAB20184-03 AAB20184-06 CAAAB20184-06 AAB20184-03
5-Fluoro-2-(trifluoromethyl)benzyl alcohol 97%
5-Fluoro-2-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
MW: 194.13 g/mol
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00236286
CAS Number: 238742-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR