2-Fluoro-5-(trifluoromethyl)benzonitrile 97%

Warning

Synonyms: 3-Cyano-4-fluorobenzotrifluoride2-Fluoro-5-trifluoromethylbenzonitrile

B20163-06 B20163-03 B20163-03
AAAB20163-03EA 55.34 USD
CAAAB20163-03 AAAB20163-03 AAAB20163-06
2-Fluoro-5-(trifluoromethyl)benzonitrile 97%
2-Fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 185…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00061280
CAS Number: 4088-84-0
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR