2,3,4-Trifluorophenol 98%

Danger

B20142-06 B20142-06
AAB20142-06EA 144.55 USD
AAB20142-06 CAAAB20142-06
2,3,4-Trifluorophenol 98%
2,3,4-Trifluorophenol
Formula: C₆H₃F₃O
MW: 148.08 g/mol
Boiling Pt: 68…70 °C/43 mm
Melting Pt: 30…34 °C
Density: 1.460
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042426
CAS Number: 2822-41-5
UN: 2925
ADR: "4.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR