2,3-Difluorocinnamic acid 98%

Warning

Synonyms: trans-2,3-Difluorocinnamic acid

B20105-06 B20105-03
CAAAB20105-03 CAAAB20105-06
2,3-Difluorocinnamic acid 98%
2,3-Difluorocinnamic acid
Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 198…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00061291
CAS Number: 207981-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR