2,3-Difluorobenzaldehyde 98%

Warning

B20093-03 B20093-06 B20093-14 B20093-06 B20093-03 B20093-14
AAB20093-03EA 37.62 USD
CAAAB20093-06 AAB20093-03 CAAAB20093-03 CAAAAB20093-14 AAAB20093-14 AAB20093-06
2,3-Difluorobenzaldehyde 98%
2,3-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 69…70 °C/15 mm
Density: 1.301
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010292
CAS Number: 2646-91-5
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR