2-Fluoro-5-(trifluoromethyl)phenol 97%

Warning

Synonyms: 1-Fluor-2-hydroxy-4-(trifluormethyl)benzol2-Fluoro-5-trifluoromethylphenol

B20004-03 B20004-03 B20004-06 B20004-06
AAB20004-06EA 110.22 USD
CAAAB20004-03 AAB20004-06 AAB20004-03 CAAAB20004-06
2-Fluoro-5-(trifluoromethyl)phenol 97%
2-Fluoro-5-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 146 °C
Density: 1.436
Flash Pt: 57 °C(134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061286
CAS Number: 141483-15-0
EINECS: 000-000-0
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR