1-Bromo-2,3,5-trifluorobenzene 98%

Warning

Synonyms: 2,3,5-Trifluorobromobenzene

B20000-06 B20000-03 B20000-06 B20000-03
AAB20000-03EA 41.49 USD
AAB20000-03 AAB20000-06 CAAAB20000-03 CAAAB20000-06
1-Bromo-2,3,5-trifluorobenzene 98%
1-Bromo-2,3,5-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 143 °C
Density: 1.777
Flash Pt: 54 °C(129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012232
CAS Number: 133739-70-5
EINECS: 000-000-0
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR