α-Bromo-3,4-difluorotoluene 98%

Danger

Synonyms: 3,4-Difluorobenzyl bromide

A19965-09 A19965-09 A19965-18
AAA19965-09EA 120.65 USD
CAAAA19965-09 CAAAAA19965-18 AAA19965-09
α-Bromo-3,4-difluorotoluene 98%
α-Bromo-3,4-difluorotoluene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 59…61 °C/3.5 mm
Density: 1.618
Flash Pt: 79 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009889
CAS Number: 85118-01-0
EINECS: 285-653-1
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR