3-Fluoro-5-(trifluoromethyl)benzonitrile 97%

Warning

Synonyms: 3-Cyano-5-fluorobenzotrifluoride

A19964-06 A19964-06 A19964-03 A19964-03
AAA19964-03EA 60.13 USD
CAAAA19964-06 CAAAA19964-03 AAA19964-03 AAA19964-06
3-Fluoro-5-(trifluoromethyl)benzonitrile 97%
3-Fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.35
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061282
CAS Number: 149793-69-1
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR