2,3-Difluorobenzophenone 97%

Warning

A19951-06 A19951-03 A19951-06 A19951-03
AAA19951-06EA 109.44 USD
CAAAA19951-06 CAAAA19951-03 AAA19951-06 AAA19951-03
2,3-Difluorobenzophenone 97%
2,3-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 176…178 °C/16 mm
Storage Temperature: Ambient
MDL Number: MFCD00061139
CAS Number: 208173-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR