Methyl-2-chloro-5-nitrobenzoate 97%

Warning

Synonyms: 2-Chloro-5-nitrobenzoic acid methyl ester

A19913-88 A19913-09 A19913-88 A19913-09
AAA19913-09EA 97.5 USD
CAAAA19913-88 AAA19913-09 AAA19913-88 CAAAA19913-09
Methyl-2-chloro-5-nitrobenzoate 97%
Methyl 2-chloro-5-nitrobenzoate
Formula: C₈H₆ClNO₄
MW: 215.59 g/mol
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00184587
CAS Number: 6307-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR