2,3,6-Trifluorobenzyl bromide 98%

Danger

Synonyms: α-Bromo-2,3,6-trifluorotoluene

A19909-03 A19909-06 A19909-06 A19909-03
AAA19909-03EA 60 USD
AAA19909-03 CAAAA19909-06 CAAAA19909-03 AAA19909-06
2,3,6-Trifluorobenzyl bromide 98%
2,3,6-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 114 °C (dec.)
Density: 1.718
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061208
CAS Number: 151412-02-1
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR