3,5-Difluorobenzamide 97%

Warning

A19904-06 A19904-03 A19904-03 A19904-06
AAA19904-03EA 29.73 USD
CAAAA19904-03 AAA19904-03 AAA19904-06 CAAAA19904-06
3,5-Difluorobenzamide 97%
3,5-Difluorobenzamide
Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00061138
CAS Number: 132980-99-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR