3-Chloro-α,α,α-trifluorotoluene 98%

Warning

Synonyms: 1-Chloro-3-(trifluoromethyl)benzene 3-Chlorobenzotrifluoride

A19854-22 A19854-22
AAA19854-22EA 153.42 USD
AAA19854-22 CAAAA19854-22
3-Chloro-α,α,α-trifluorotoluene 98%
3-Chloro-α,α,α-trifluorotoluene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 137…138 °C
Melting Pt: –56 °C
Density: 1.331
Flash Pt: 38 °C(100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000594
CAS Number: 98-15-7
EINECS: 202-642-9
UN: 2234
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR