2-Octynoic acid ethyl ester 98%

Synonyms: Ethyl 2-octynoate

A19784-14 A19784-22 A19784-14 A19784-06 A19784-22 A19784-06
AAA19784-06EA 42.85 USD
CAAAA19784-22 AAA19784-06 CAAAA19784-14 AAA19784-14 CAAAA19784-06 AAA19784-22
2-Octynoic acid ethyl ester 98%
2-Octynoic acid ethyl ester
Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
Boiling Pt: 85 °C/6 mm
Density: 0,910
Flash Pt: 99 °C(210 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00015286
CAS Number: 10519-20-7
EINECS: 234-058-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR