Pentafluoropyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,3,4,5,6-Pentafluoropyridine
A19656-06
A19656-14
A19656-06
A19656-14
AAA19656-06EA
56.22
USD
CAAAA19656-06
CAAAA19656-14
AAA19656-06
AAA19656-14
Pentafluoropyridine 99%
Pentafluoropyridine
Formula:
C₅F₅N MW: 169.05 g/mol Boiling Pt: 83…85 °C Melting Pt: –41 °C Density: 1.618 Flash Pt: 23 °C(73 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006225 CAS Number: 700-16-3 EINECS: 211-839-9 UN: 2924 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...