1-Tetradecanol 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Tetradecyl alcohol
A19638-22
A19638-36
A19638-0E
A19638-22
A19638-36
A19638-0E
AAA19638-0EEA
146.99
USD
AAA19638-0E
CAAAAA19638-22
AAAA19638-22
CAAAA19638-36
CAAAA19638-0E
AAA19638-36
1-Tetradecanol 97+%
1-Tetradecanol
Formula:
H₃C(CH₂)₁₂CH₂OH MW: 214.39 g/mol Boiling Pt: 289 °C Melting Pt: 37…40 °C Density: 0.823 Flash Pt: 141 °C(285 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004757 CAS Number: 112-72-1 EINECS: 204-000-3 UN: 3077 ADR: "9",III Merck Index: 13,06361 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...