4-(Trifluoromethoxy)benzoyl chloride 97%

Danger

Synonyms: 4-(Trifluoromethoxy)benzene-1-carbonyl chloride

A19452-14 A19452-06 A19452-03 A19452-14 A19452-03 A19452-06
AAA19452-06EA 105.78 USD
CAAAA19452-03 CAAAA19452-06 CAAAAA19452-14 AAA19452-06 AAA19452-03 AAAA19452-14
4-(Trifluoromethoxy)benzoyl chloride 97%
4-(Trifluoromethoxy)benzoyl chloride
Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Boiling Pt: 55…56 °C/2 mm
Density: 1.425
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052329
CAS Number: 36823-88-8
EINECS: 000-000-0
UN: 2920
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR