1,3-Cyclohexadiene 96% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclohexa-1,3-diene

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.01%

A19394-06 A19394-14 A19394-06 A19394-14
AAA19394-06EA 64.08 USD
CAAAA19394-14 AAA19394-06 CAAAA19394-06 AAA19394-14
1,3-Cyclohexadiene 96% stabilized
1,3-Cyclohexadiene
Formula: C₆H₈
MW: 80.13 g/mol
Boiling Pt: 79…81 °C
Melting Pt: –89 °C
Density: 0.847
Flash Pt: -8 °C(-144 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001532
CAS Number: 592-57-4
EINECS: 209-764-1
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR