Methyl-5-chloro-2-nitrobenzoate 98%

Warning

Synonyms: 5-Chloro-2-nitrobenzoic acid methyl ester

A19098-14 A19098-03 A19098-06 A19098-14 A19098-03 A19098-06
AAA19098-03EA 28.97 USD
AAA19098-03 AAA19098-14 CAAAA19098-06 CAAAA19098-14 CAAAA19098-03 AAA19098-06
Methyl-5-chloro-2-nitrobenzoate 98%
Methyl 5-chloro-2-nitrobenzoate
Formula: C₈H₆ClNO₄
MW: 215.59 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 48…50 °C
Density: 1,426 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00272139
CAS Number: 51282-49-6
EINECS: 257-107-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR